Learn More
Eisco™ Flask Electroscope with Aluminum Leaf
Catalog No. S23684
Change view
Click to view available options
Aluminum Leaf Electroscope
Catalog No.
S23684 Aluminum Leaf Electroscope
1 options
Catalog No. S23684 Supplier Eisco™ Supplier No. PH0912
An aluminum leaf electroscope for observation of polarity through electric charge detection.
  • Brass rod and aluminum lead
  • Glass conical flask
  • 7.5 in. height with 3.5 in diameter
  • Detect electric charges and determine polarity
Specifications
For Use With (Application) Measure potential difference between the leaf and the base
Includes A glass conical flask, brass rod with disc and aluminum leaf attached supported rubber cork
Product Type Aluminum Leaf Electroscope, Flask Type