missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) oxide, Premion™, 99.99% (metals basis), Au 88.6% min

Catalog No. p-7048940
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1216101 0.25 g
AA1216103 1 g
AA1216106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1216101 Supplier Thermo Scientific Chemicals Supplier No. 01216101

CAS: 1303-58-8 | Au2O3 | 441.93 g/mol

It is used in aerospace and electrochemical applications such as fuel cells in which they exhibit ionic conductivity. It acts as strong reducing agent in redox reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1303-58-8
Molecular Formula Au2O3
Molecular Weight (g/mol) 441.93
MDL Number MFCD00014173
InChI Key DDYSHSNGZNCTKB-UHFFFAOYSA-N
Synonym digold trioxide, unii-lw5y686rup, gold iii oxide, gold 3+ ; oxygen 2-, lw5y686rup, gold trioxide, gold oxide, gold sesquioxide, gold oxide,, gold iii oxide, premion
PubChem CID 164805
IUPAC Name gold(3+);oxygen(2-)
SMILES [O-2].[O-2].[O-2].[Au+3].[Au+3]

Specifications

Melting Point 150°C (decomposition)
Color Brown
Physical Form Powder
Quantity 0.25 g
Merck Index 14,4524
Solubility Information Insoluble in water; Soluble in HCl,HNO3and in NaCN solution.
Formula Weight 441.93
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Au 88.6% min.
Odor Odorless
Assay (metals basis)
Chemical Name or Material Gold(III) oxide
Show More Show Less
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 215-122-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only