missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) hydroxide, Au 79% min

Catalog No. p-7048939
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1215903 1 g
AA1215906 5 g
AA1215909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1215903 Supplier Thermo Scientific Chemicals Supplier No. 01215903

Gold plating solutions, porcelain decoration | CAS: 1303-52-2 | AuH6O3 | 251.01 g/mol

Gold(III) hydroxide is used in medicine, porcelain making, gold plating, and daguerrotypes. Used for preparation of gold catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1303-52-2
Molecular Formula AuH6O3
Molecular Weight (g/mol) 251.01
MDL Number MFCD00046173
InChI Key PPOYUERUQZXZBE-UHFFFAOYSA-N
Synonym gold iii hydroxide, gold trihydroxide, gold hydroxide au oh 3, auh3o3, gold hydroxide, au oh 3, gold iii hydroxide, au
PubChem CID 102103
SMILES O.O.O.[Au]

Specifications

Physical Form Powder
Quantity 1 g
Merck Index 14,4523
Solubility Information Insoluble in water.
IUPAC Name gold;trihydrate
Formula Weight 247.99
Concentration or Composition (by Analyte or Components) Au 79% min.
Odor Odorless
Sensitivity Light sensitive
Chemical Name or Material Gold(III) hydroxide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-120-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only