missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) chloride trihydrate, Honeywell Fluka™

Catalog No. 6000627
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000627 1 g
6000628 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000627 Supplier Honeywell Chemicals Supplier No. G40221G
Only null left

ACS Reagent,≥49.0% Au basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
MDL Number MFCD00149904
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Quantity 1 g
Linear Formula HAuCl4 · 3H2O
UN Number UN3260
Formula Weight 393.83g/mol
Percent Purity ≥49% (Au basis)
Chemical Name or Material Gold(III) chloride trihydrate
Hazard Statement P280-P305 + P351 + P338-P310
RTECSNumber MD5428000