missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Gold(III) chloride trihydrate

Catalog No. p-200140773
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
5209185G 5 g
52091825G 25 g
5209181G 1 g
3 options

Catalog No. 5209185G

Supplier: Sigma Aldrich 5209185G

Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The embryotoxicity of gold salt (HAuCl4 -+ 3H2O) is weaker than the embryotoxicities of a number of other nanoparticles used infor biomedical applications.Gold(III) chloride trihydrate can be used in the fabrication of colorimetric aptasensor for staphylococcus aureus. It can be reduced with polygala tenuifolia roots (green reducing agent) to produce gold nanoparticles (AuNPs).

Specifications

CAS 16961-25-4
Molecular Formula HAuCl4 -+ 3H2O
Linear Formula HAuCl4 -+ 3H2O
Quantity 5 g
MDL Number MFCD00149904
Synonym Hydrogen tetrachloroaurate(III); Tetrachloroauric(III) acid
Molecular Weight (g/mol) 393.83
Percent Purity ≥99.9% trace metals basis