missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Gold(III) chloride hydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$1469.88 - $1469.88
Specifications
| CAS | 27988-77-8 |
|---|---|
| Density | 3.9 g/mL (at 25°C) |
| Molecular Formula | HAuCl4 -+ xH2O |
| Linear Formula | HAuCl4 -+ aq |
| MDL Number | MFCD00149903 |
Description
The structure of gold chloride is monoclinic in nature. It sublimes at elevated temperatures.It is used in the preparation of stable nanosized and underivatized colloidal gold particles in solution.Specifications
| 27988-77-8 | |
| HAuCl4 -+ xH2O | |
| MFCD00149903 | |
| 339.79 |
| 3.9 g/mL (at 25°C) | |
| HAuCl4 -+ aq | |
| Auric chloride; Chloroauric acid; Hydrogen tetrachloroaurate(III) hydrate; Tetrachloroauric(III) acid | |
| 99.995% trace metals basis |