missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) bromide, Premion™, 99.99% (metals basis), Au 44.6% min, Thermo Scientific Chemicals

Catalog No. p-7048941
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1216203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1216203 Supplier Thermo Scientific Chemicals Supplier No. 01216203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10294-28-7 | Molecular Formula: AuBr3H3 | Molecular Weight: 439.70 g/mol

Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10294-28-7
Molecular Formula AuBr3H3
Molecular Weight (g/mol) 439.70
MDL Number MFCD00014171
InChI Key KJCSJLXVPHOFJC-UHFFFAOYSA-N
IUPAC Name gold(III) bromide
SMILES Br.Br.Br.[Au]

Specifications

Melting Point ∼160°C (decomposition)
Physical Form Granules
Quantity 1 g
UN Number UN3260
Solubility Information Slightly soluble in water.
Formula Weight 436.69
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Au 44.6% min.
Assay (metals basis)
Chemical Name or Material Gold(III) bromide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 233-654-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only