missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) bromide, 99%

Catalog No. AAA1951403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1951403 1 g
AAA1951406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1951403 Supplier Thermo Scientific Chemicals Supplier No. A1951403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10294-28-7 | AuBr3H3 | 439.70 g/mol

Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 10294-28-7
Molecular Formula AuBr3H3
Molecular Weight (g/mol) 439.70
MDL Number MFCD00014171
InChI Key KJCSJLXVPHOFJC-UHFFFAOYSA-N
IUPAC Name gold(III) bromide
SMILES Br.Br.Br.[Au]

Specifications

Melting Point ∼160°C (decomposition)
Quantity 1 g
UN Number UN3260
Solubility Information Slightly soluble in water.
Formula Weight 436.69
Percent Purity 99%
Sensitivity Hygroscopic
Chemical Name or Material Gold(III) bromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 233-654-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only