missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) chloride, 99.9% (metals basis)

CAS: 10294-29-8 | AuCl | 232.42 g/mol

$549.01 - $549.01

Chemical Identifiers

CAS 10294-29-8
Molecular Formula AuCl
Molecular Weight (g/mol) 232.42
MDL Number MFCD00046175
InChI Key FDWREHZXQUYJFJ-UHFFFAOYSA-M
Synonym gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade
PubChem CID 27366
ChEBI CHEBI:30078
IUPAC Name chlorogold
SMILES [Cl-].[Au+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4336503
View Documents
Thermo Scientific Chemicals
04336503
1 g
Each for $549.01
Only null left
 
Description

Description

Gold(I) chloride is used as a starting material for the preparation of Phosphine ligated gold(I) complexes, which is used as catalysts in synthetic chemistry. It is also used to prepare metallic gold and potassium auric bromide and potassium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10294-29-8
232.42
FDWREHZXQUYJFJ-UHFFFAOYSA-M
27366
chlorogold
AuCl
MFCD00046175
gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade
CHEBI:30078
[Cl-].[Au+]

Specifications

289°C (decomposition)
Powder
AuCl
UN3260
gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade
FDWREHZXQUYJFJ-UHFFFAOYSA-M
chlorogold
27366
232.42
Odorless
7.57 g/mL
(metals basis)
10294-29-8
1 g
MFCD00046175
14,4515
Soluble in hydrochloric acid,hydrobromic acid and organic solvents. Slightly soluble in water.
[Cl-].[Au+]
232.42
CHEBI:30078
99.9%
Moisture sensitive
Gold (I) chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P234-P260-P264b-P272-P280-P301+P330+P331-P302+P352-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H317-H500

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 233-655-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only