missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Chemical Gold Chloride, Trihydrate, ACS, 49%, Spectrum™ Chemical

Catalog No. 18610751
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610751 1 g Amber Glass Bottle
18610752 5 g Amber Glass Bottle
18610753 10 g Amber Glass Bottle
18610754 25 g Amber Glass Bottle
18610755 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610751 Supplier Spectrum™ Chemical Supplier No. G10251GM
Get Up to $500 in Chemicals with Purchase
View all promotions
  • Grade: ACS
  • Assay (Au): 49.0%
  • Molecular Formula: HAuCl4·3H2O
  • Physical State: Solid
  • Appearance: Crystalline
  • Color: Yellow to red
  • Solubility: Soluble in water, ether, and ethanol
  • Melting Point: 254°C
  • Relative Density: 3.9
  • Storage: Light sensitive, deliquescent; keep tightly closed in light-resistant containers

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Melting Point 254°C
Color Yellow to Red
CAS Max % 1
Physical Form Solid
Quantity 1 g
Assay Percent Range 49.0% (Au) min
Linear Formula HAuCl4·3H2O
UN Number UN3260
Synonym Tetrachloroauric (III) Acid
Solubility Information Soluble in Water, Ether, Ethanol
Formula Weight 339.77 g/mol
Percent Purity 49%
Grade ACS
Packaging Amber Glass Bottle
Sensitivity Light Sensitive, Deliquescent
Appearance Crystalline
Density 3.9 g/cm³
Chemical Name or Material Gold Chloride
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.