Learn More
Gold Chloride, 1% (w/v), LabChem™
Supplier: LabChem LC148957
Specifications
| Gold Chloride | |
| 16961-25-4,7732-18-5 | |
| Liquid | |
| AuCl4H | |
| Soluble in water | |
| Cl.Cl[Au](Cl)Cl | |
| 339.77 | |
| 1% (w/v) | |
| Amber Glass |
| Yellow | |
| 98.9,1.1 | |
| 125 mL | |
| HAuCl4·3H2O | |
| VDLSFRRYNGEBEJ-UHFFFAOYSA-K | |
| trichlorogold hydrochloride | |
| 393.83 | |
| Passes Test | |
| Hydrogen chloride |
Chemical Identifiers
| 16961-25-4 | |
| 339.77 | |
| trichlorogold hydrochloride |
| AuCl4H | |
| VDLSFRRYNGEBEJ-UHFFFAOYSA-K | |
| Cl.Cl[Au](Cl)Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
Recommended Storage : Room Temperature