missing translation for 'onlineSavingsMsg'
Learn More

GOJO™ Scrubbing Wipes

Catalog No. 19043741
Change view
Click to view available options
Packaging Quantity:
72 Wipes/Bucket, 6 Buckets/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
19043741 72 Wipes/Bucket, 6 Buckets/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19043741 Supplier GOJO™ Supplier No. 6396 06

Wipes cean the toughest soils, no water is needed.

  • Made of durable, dual-textured material pre-moistened with a powerful cleaning solution.
  • The scrubbing side removes the toughest soils, while the smooth side quickly absorbs dirt.
  • Extra large wipes for superior cleaning capacity
  • Contain skin conditioners to help prevent dryness and irritation
  • For use anywhere heavy-duty cleaning is required; no water needed

Specifications

Product Type Sanitizing Wipe
For Use With (Application) Removes the toughest soils, while the smooth side quickly absorbs dirt
Packaging Quantity 72 Wipes/Bucket, 6 Buckets/Cs.
Size 72 count
pH Range 4.5 to 8.5