missing translation for 'onlineSavingsMsg'
Learn More

GOJO™ Cherry Gel Pumice Hand Cleaner

Catalog No. 19158231
Change view
Click to view available options
Volume (Metric):
2000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19158231 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19158231 Supplier GOJO™ Supplier No. 7290 04

Gel-style heavy-duty hand cleaner with pumice scrubbers quickly cleans heavy dirt, grease and oil

Provides a quick, easy and complete rinse to leave skin feeling refreshed.

  • pH balanced to promote skin comfort
      Pleasant cherry fragrance eliminates soil odors
    • Clings to hands for easy cleaning; doesn't fall off your hands into the sink
    • Easy to use and spread across hands and arms

Specifications

Product Type Pumice Hand Cleaner
Color Red
Form Gel
Composition Petroleum Distillates, Oleic Acid
For Use With (Application) Quickly cleans heavy dirt, grease and oil
Packaging Refill
pH Range 6.5 to 8
Quantity 4 Pk.
Volume (English) 0.52 gal.
Volume (Metric) 2000 mL