Learn More
Moldex™ Goin' Green™ PlugStation™ Ear Plug Dispenser
Catalog No. 191600159
Change view
Click to view available options
Holds:
250 Pairs
Catalog No. Holds
191600159 250 Pairs
1 options
Catalog No. 191600159 Supplier Moldex™ Supplier No. 6646
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Goin' Green™ PlugStation™ dispensers are pre-filled with ear plugs and are convenient for any work environment. Easy access, wall-mountable unit will help ensure compliance. When the PlugStation is empty, simply dispose of it and hang a new one.
  • Holds Goin' Green™ disposable ear plugs
  • PlugStations are sealed to ensure ear plugs stay clean and sanitary
  • High visibility to increase compliance
  • Dispenses one pair at a time to eliminate wasteful grabbing
  • Includes mounting bracket
  • Available in two sizes - Small 250 Pair/Dispenser or Large 500 Pair/Dispenser
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Easy Access to Hearing Protection for Workers and Facility Visitors in Reception Areas, Break Rooms, Workstations, and Other Noisy Environments.
Holds 250 Pairs
Pairs per Package 250 Pairs/Dispenser
Plug Material Foam
Plug Color Green
Noise Reduction Rating (NRR) 33 dB
Corded Uncorded
Plug Shape Bullet
Disposable Single-use
Material PVC-free Plastic
Mounting Wall Mountable
Includes Mounting Hardware
Show More Show Less