Learn More
3M™ GoggleGear™ 6000 Series Scotchgard™ Goggles
Catalog No. p-7159941
Change view
Click to view available options
Frame Color:
Black
Lens Tint:
Clear
Headband Material:
Nylon
Catalog No. Frame Color Lens Tint Headband Material
19071096 Black Clear Nylon
1 options
Catalog No. 19071096 Supplier 3M™ Supplier No. 7100191924
Only null left
3M™ Scotchgard™ Anti-Fog Coating helps to increase eyewear performance in wet and steamy environments.
  • Indirect venting allows air to circulate and helps reduce fogging
  • D3 (droplet and splash) and D4 (dust) rated
  • Scotchgard™ Anti-fog coating for fog and abrasion resistance
  • Polycarbonate lens absorbs 99.9% UVA & UVB and meet the requirements of ANSI Z87.1- 2015 Standard as an Impact Rated spectacle
  • 3M Catalog# GG6001NSGAF-BLU, GG6001SGAF-BLK, GG6001SGAF-GRN, GG6001SGAF-RED, GG6002SGAF-BLK
Specifications
Lens Coating Anti-fog, UV-treated
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Product Type Safety Goggles
For Use With (Application) Food and Beverage Manufacturing, Chemical Handling, Primary Metals, Painting, Oil and Gas
Frame Color Black
Headband Material Nylon
No. per Case 10/Cs.
Model GG6001SGAF-BLK
Size Universal
Ventilation Details Indirect venting
Material Polycarbonate
Lens Material Polycarbonate
UV Protection UV400
Show More Show Less