missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat IgG Isotype Control

Catalog No. 026202
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
026202 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 026202 Supplier Invitrogen™ Supplier No. 026202
Only null left

Goat Polyclonal Isotype Control

Goat IgG is purified from pooled normal goat serum using various methods. Purity is verified by SDS-PAGE analysis. The concentration of 5 mg/mL based on an E (1%) = 14.0 at 280 nm.

Goat IgG, also known as goat immunoglobulin G, is an antibody that is produced by the immune system of goats in response to the presence of antigens. IgG is a type of immunoglobulin that plays a role in the recognition and neutralization of pathogens, such as bacteria and viruses. It binds to specific antigens and triggers various immune responses, including phagocytosis and complement activation. Goat IgG can be used in research to study the immune response to pathogens, as well as to develop diagnostic tests and therapeutic interventions for infectious diseases.

Specifications

Antigen Goat IgG
Applications Control
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias Immunoglobulin G; ImmunoglobulinG
Host Species Goat
Purification Method Purified
Quantity 10 mg
Regulatory Status RUO
Target Species Not Applicable
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Isotype Control
Form Liquid
Isotype IgG
Show More Show Less