Learn More
SignalChem Biotech YES1 (T348I), Active 50ug
Catalog No. 502572537
Encompass
Full-length recombinant human YES1 (T348I) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502572537 Supplier SignalChem Biotech Supplier No. Y0112BG50
Full-length recombinant human YES1 (T348I) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.