Learn More
ACROBiosystems Inc ACROBiosystems Mouse Glypican 1 / GPC1 Protein, His Tag
Catalog No. 501956772
Encompass
Recombinant Protein;1MG;Mouse Glypican 1, His Tag (GP1-M52H7) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 529 (Accession # Q9QZF2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.5 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 1 g/mL (100 L/well) can bind Mouse Glypican 1, His Tag (Cat. No. GP1-M52H7) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 501956772 Supplier ACROBiosystems Inc Supplier No. GP1M52H71MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;1MG;Mouse Glypican 1, His Tag (GP1-M52H7) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 529 (Accession # Q9QZF2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.5 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 1 g/mL (100 L/well) can bind Mouse Glypican 1, His Tag (Cat. No. GP1-M52H7) with a linear range of 5-78 ng/mL (QC tested).