missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glyphosate

Catalog No. AC466180010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466180010 1 g
AC466180050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466180010 Supplier Thermo Scientific Chemicals Supplier No. 466180010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1071-83-6 | C3H8NO5P | 169.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1071-83-6
Molecular Formula C3H8NO5P
Molecular Weight (g/mol) 169.07
MDL Number MFCD00055350
InChI Key XDDAORKBJWWYJS-UHFFFAOYSA-N
IUPAC Name 2-[(phosphonomethyl)amino]acetic acid
SMILES OC(=O)CNCP(O)(O)=O

Specifications

Melting Point 230.0°C
Solubility Information Solubility aq. soln.: soluble. Other solubilities: insoluble in most organic solvents
Quantity 1 g
Formula Weight 169.07
Chemical Name or Material Glyphosate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes serious eye damage.

Toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 213-997-4

RUO – Research Use Only