missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyoxylic acid monohydrate, 98%, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 48.0°C to 52.0°C |
| Color | White to Yellow |
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Plastic bottle |
| Linear Formula | HCOCO2H·H2O |
| Quantity | 25 g |
| Solubility Information | Solubility in water: freely soluble. |
| Formula Weight | 92.06 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes serious eye damage. May cause an allergic skin reaction. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear eye protection/face protecti |
RUO – Research Use Only