missing translation for 'onlineSavingsMsg'
Learn More

Glyoxylic acid monohydrate, 97%

Catalog No. AAA1605814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1605814 25 g
AAA1605822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1605814 Supplier Thermo Scientific Chemicals Supplier No. A1605814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 563-96-2 | C2H4O4 | 92.05 g/mol

Glyoxylic acid is a reagent for sulfinylmaleate synthesis. It is one of the chemicals used in the Hopkins Cole reaction. It is involved to check the presence of tryptophan in proteins. It is condensed with urea and 1,2-diaminobenzene to form heterocycles. It also undergoes Friedel-Crafts and cyclocondensation reactions to form bis(pentamethylphenyl) acetic acid and a beta-carboline respectively. It is employed in the synthesis of a sulfinylmaleate, which serves as an efficient dienophile for enantioselective Diels-Alder cycloadditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glyoxylic acid is a reagent for sulfinylmaleate synthesis. It is one of the chemicals used in the Hopkins Cole reaction. It is involved to check the presence of tryptophan in proteins. It is condensed with urea and 1,2-diaminobenzene to form heterocycles. It also undergoes Friedel-Crafts and cyclocondensation reactions to form bis(pentamethylphenyl) acetic acid and a beta-carboline respectively. It is employed in the synthesis of a sulfinylmaleate, which serves as an efficient dienophile for enantioselective Diels-Alder cycloadditions.

Solubility
Soluble in water, dimethyl sulfoxide and alcohol. Insoluble in diethyl ether and hydrocarbons.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible withbases, oxidizing agents and reducing agents.

Chemical Identifiers

CAS 563-96-2
Molecular Formula C2H4O4
Molecular Weight (g/mol) 92.05
MDL Number MFCD00149497
InChI Key MOOYVEVEDVVKGD-UHFFFAOYSA-N
Synonym glyoxylic acid monohydrate, glyoxylic acid hydrate, 2-oxoacetic acid hydrate, oxaldehydic acid hydrate, glyoxalic acid monohydrate, glyoxalate, glyoxylate hydrate, glyoxalic acid hydrate, oxoacetate monohydrate, glyoxylic acid-hydrate, oxoacetic acid hydrate
PubChem CID 15620607
IUPAC Name oxaldehydic acid;hydrate
SMILES C(=O)C(=O)O.O

Specifications

Melting Point 48°C to 52°C
Boiling Point 100°C
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3261
Beilstein 969535
Sensitivity Hygroscopic
Merck Index 14,4511
Solubility Information Soluble in water,dimethyl sulfoxide and alcohol. Insoluble in diethyl ether and hydrocarbons.
Formula Weight 92.06 (74.04 Anhydrous)
Percent Purity 97%
Chemical Name or Material Glyoxylic acid monohydrate
Show More Show Less
Hazard Category H314-H317-H500
Hazard Statement GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 206-058-5
RTECSNumber MD4550000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only