missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Glyoxylic acid monohydrate
Description
Glyoxylic acid monohydrate
Specifications
Specifications
| CAS | 563-96-2 |
| Melting Point | 49°C to 52°C (lit.) |
| Boiling Point | 100°C (lit.) |
| Molecular Formula | C2H2O3 -+ H2O |
| Linear Formula | HCOCO2H -+ H2O |
| MDL Number | MFCD00149497 |
| Quantity | 10 g |
| Synonym | Formylformic acid; Oxoethanoic acid |
| Molecular Weight (g/mol) | 92.05 |
| Percent Purity | 98% |