missing translation for 'onlineSavingsMsg'
Learn More

Glyoxylic Acid (ca. 50% in Water, ca. 9mol/L), TCI America™

Catalog No. G036625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G036625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G036625ML Supplier TCI America Supplier No. G036625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 298-12-4
Molecular Formula C2H2O3
Molecular Weight (g/mol) 74.035
MDL Number MFCD00006958
InChI Key HHLFWLYXYJOTON-UHFFFAOYSA-N
Synonym glyoxylic acid, 2-oxoacetic acid, oxoacetic acid, oxoethanoic acid, glyoxalic acid, formylformic acid, acetic acid, oxo, oxalaldehydic acid, glyoxylate, alpha-ketoacetic acid
PubChem CID 760
ChEBI CHEBI:16891
IUPAC Name oxaldehydic acid
SMILES C(=O)C(=O)O

Specifications

Color Yellow
Quantity 25 mL
UN Number 3265
Formula Weight 74.04
Physical Form Liquid
Chemical Name or Material Glyoxylic Acid (ca. 50% in Water, ca. 9mol/L)
EINECSNumber (2)-1471
RTECSNumber MD4550000
TSCA Yes