missing translation for 'onlineSavingsMsg'
Learn More

Glyoxalbis(2-hydroxyanil), 97%

CAS: 1149-16-2 | C14H12N2O2 | 240.26 g/mol

$178.41 - $561.28

Chemical Identifiers

CAS 1149-16-2
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00002183
InChI Key OMJNMRJMNUREOJ-KOBPNRPCSA-N
Synonym glyoxal bis 2-hydroxyanil, glyoxalbis 2-hydroxyanil, glyoxal bis o-hydroxyanil, glyoxal bis 2-hydroxyaniline, glyoxylidenebis 2-hydroxyaniline, unii-e2aw5xx09u, gbha, 2,2'-ethanediylidenedinitrilo bisphenol, phenol, 2,2'-1,2-ethanediylidenedinitrilo bis, 2,2'-ethanediylidenedinitrilo diphenol
PubChem CID 5356911
SMILES OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120160100
SDS View Documents
Thermo Scientific Chemicals
120160100
10 g Glass bottle
Each for $178.41
Only null left
 
AC120160500
SDS View Documents
Thermo Scientific Chemicals
120160500
50 g Glass bottle
Each for $561.28
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

1149-16-2
240.26
OMJNMRJMNUREOJ-KOBPNRPCSA-N
5356911
C14H12N2O2
MFCD00002183
glyoxal bis 2-hydroxyanil, glyoxalbis 2-hydroxyanil, glyoxal bis o-hydroxyanil, glyoxal bis 2-hydroxyaniline, glyoxylidenebis 2-hydroxyaniline, unii-e2aw5xx09u, gbha, 2,2'-ethanediylidenedinitrilo bisphenol, phenol, 2,2'-1,2-ethanediylidenedinitrilo bis, 2,2'-ethanediylidenedinitrilo diphenol
OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O

Specifications

1149-16-2
Beige
Authentic
Glass bottle
MFCD00002183
glyoxal bis 2-hydroxyanil, glyoxalbis 2-hydroxyanil, glyoxal bis o-hydroxyanil, glyoxal bis 2-hydroxyaniline, glyoxylidenebis 2-hydroxyaniline, unii-e2aw5xx09u, gbha, 2,2'-ethanediylidenedinitrilo bisphenol, phenol, 2,2'-1,2-ethanediylidenedinitrilo bis, 2,2'-ethanediylidenedinitrilo diphenol
OMJNMRJMNUREOJ-KOBPNRPCSA-N
6-{[(1E)-2-[(2-hydroxyphenyl)amino]ethenyl]imino}cyclohexa-2,4-dien-1-one
5356911
97%
Glyoxalbis(2-hydroxyanil)
201°C to 205°C
0.5% max. (105°C)
96% min. (HPLC)
C14H12N2O2
10 g
(0.5% in methanol) Clear
OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O
240.26
240.26
Fine Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-560-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only