missing translation for 'onlineSavingsMsg'
Learn More

Glyoxal Sodium Bisulfite Hydrate (contains oligomers) 97.0+%, TCI America™

Catalog No. G015425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G015425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G015425G Supplier TCI America Supplier No. G015425G

Specifications

CAS 332360-05-1
Color White
Molecular Formula C2H4Na2O8S2
MDL Number MFCD0149541
Quantity 25 g
Synonym glyoxal-sodium bisulfite monohydrate, glyoxal sodium bisulfite addition compound hydrate, 1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt hydrate, 1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt, c2h4o8s2.2na.h2o, glyoxal-sodium bisulfite monohydrate mi, glyoxal bis sodium hydrogen sulfite adduct hydrate, glyoxal sodium bisulfite addition compound hydrate,98, disodium 1,2-dihydroxy-1,2-ethanedisulfonate monohydrate, disodium 1,2-dihydroxyethane-1,2-disulfonate hydrate
InChI Key BXUKAXFDABMVND-UHFFFAOYNA-L
SMILES [Na+].[Na+].OC(C(O)S([O-])(=O)=O)S([O-])(=O)=O
IUPAC Name disodium 1,2-dihydroxyethane-1,2-disulfonate
Molecular Weight (g/mol) 266.15
PubChem CID 16211366
Formula Weight 266.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glyoxal Sodium Bisulfite Hydrate (contains oligomers)
Show More Show Less
EINECSNumber (2)-1596
TSCA Yes