missing translation for 'onlineSavingsMsg'
Learn More

Glyoxal dimethyl acetal, 60% aq. soln.

Catalog No. AAL1849022
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1849022 100 g
AAL1849036 500 g
AAL184900P 2000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1849022 Supplier Thermo Scientific Chemicals Supplier No. L1849022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51673-84-8 | C4H8O3 | 104.105 g/mol

Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 51673-84-8
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00134410
InChI Key OGFKTAMJLKHRAZ-UHFFFAOYSA-N
Synonym acetaldehyde, dimethoxy, dimethoxyacetaldehyde, dimethoxy acetaldehyde, glyoxal dimethylacetal, glyoxal dimethyl acetal, 60 wt. % in h2o, acetaldehyde, 2,2-dimethoxy, 2,2-dimethoxyethanal, glyoxaldimethylacetal, dimethoxyethanal
PubChem CID 162650
IUPAC Name 2,2-dimethoxyacetaldehyde
SMILES COC(C=O)OC

Specifications

Density 1.15
Boiling Point 100°C to 102°C
Flash Point 102°C (216°F)
Refractive Index 1.414
Quantity 100 g
Beilstein 1850744
Solubility Information Fully miscible in water.
Formula Weight 104.11
Concentration or Composition (by Analyte or Components) 60% aq. soln.
Chemical Name or Material Glyoxal dimethyl acetal
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only