missing translation for 'onlineSavingsMsg'
Learn More

Glyoxal Bis(2-hydroxyanil) 98.0+%, TCI America™

$130.92 - $342.69

Chemical Identifiers

CAS 1149-16-2
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00002183
InChI Key OMJNMRJMNUREOJ-KOBPNRPCSA-N
Synonym glyoxal bis 2-hydroxyanil, glyoxalbis 2-hydroxyanil, glyoxal bis o-hydroxyanil, glyoxal bis 2-hydroxyaniline, glyoxylidenebis 2-hydroxyaniline, unii-e2aw5xx09u, gbha, 2,2'-ethanediylidenedinitrilo bisphenol, phenol, 2,2'-1,2-ethanediylidenedinitrilo bis, 2,2'-ethanediylidenedinitrilo diphenol
PubChem CID 5356911
IUPAC Name 6-{[(1E)-2-[(2-hydroxyphenyl)amino]ethenyl]imino}cyclohexa-2,4-dien-1-one
SMILES OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
G01535G
View Documents
TCI America
G01535G
5 g
Each for $130.92
Only null left
 
G015325G
View Documents
TCI America
G015325G
25 g
Each for $342.69
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1149-16-2
240.26
OMJNMRJMNUREOJ-KOBPNRPCSA-N
5356911
OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O
C14H12N2O2
MFCD00002183
glyoxal bis 2-hydroxyanil, glyoxalbis 2-hydroxyanil, glyoxal bis o-hydroxyanil, glyoxal bis 2-hydroxyaniline, glyoxylidenebis 2-hydroxyaniline, unii-e2aw5xx09u, gbha, 2,2'-ethanediylidenedinitrilo bisphenol, phenol, 2,2'-1,2-ethanediylidenedinitrilo bis, 2,2'-ethanediylidenedinitrilo diphenol
6-{[(1E)-2-[(2-hydroxyphenyl)amino]ethenyl]imino}cyclohexa-2,4-dien-1-one

Specifications

1149-16-2
White-Yellow
MFCD00002183
glyoxal bis 2-hydroxyanil, glyoxalbis 2-hydroxyanil, glyoxal bis o-hydroxyanil, glyoxal bis 2-hydroxyaniline, glyoxylidenebis 2-hydroxyaniline, unii-e2aw5xx09u, gbha, 2,2'-ethanediylidenedinitrilo bisphenol, phenol, 2,2'-1,2-ethanediylidenedinitrilo bis, 2,2'-ethanediylidenedinitrilo diphenol
OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O
240.26
240.26
Crystalline Powder
205°C
C14H12N2O2
5 g
OMJNMRJMNUREOJ-KOBPNRPCSA-N
6-{[(1E)-2-[(2-hydroxyphenyl)amino]ethenyl]imino}cyclohexa-2,4-dien-1-one
5356911
≥98.0% (N)
Glyoxal Bis(2-hydroxyanil)
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'