missing translation for 'onlineSavingsMsg'
Learn More

Glyoxal Bis(2-hydroxyanil) 98.0+%, TCI America™

Catalog No. G01535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01535G 5 g
G015325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G01535G Supplier TCI America Supplier No. G01535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1149-16-2
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00002183
InChI Key OMJNMRJMNUREOJ-KOBPNRPCSA-N
Synonym glyoxal bis 2-hydroxyanil, glyoxalbis 2-hydroxyanil, glyoxal bis o-hydroxyanil, glyoxal bis 2-hydroxyaniline, glyoxylidenebis 2-hydroxyaniline, unii-e2aw5xx09u, gbha, 2,2'-ethanediylidenedinitrilo bisphenol, phenol, 2,2'-1,2-ethanediylidenedinitrilo bis, 2,2'-ethanediylidenedinitrilo diphenol
PubChem CID 5356911
IUPAC Name 6-{[(1E)-2-[(2-hydroxyphenyl)amino]ethenyl]imino}cyclohexa-2,4-dien-1-one
SMILES OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O

Specifications

Melting Point 205°C
Color White-Yellow
Quantity 5 g
Formula Weight 240.26
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Glyoxal Bis(2-hydroxyanil)