missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyoxal-bis(2-hydroxyanil), ≥97.0% (from N), Honeywell Fluka™
Indicator for metal titration
$1079.36 - $5728.02
Chemical Identifiers
| CAS | 1149-16-2 |
|---|---|
| Molecular Formula | C14H12N2O2 |
| Molecular Weight (g/mol) | 240.26 |
| MDL Number | MFCD00002183 |
| InChI Key | OMJNMRJMNUREOJ-KOBPNRPCSA-N |
| PubChem CID | 5356911 |
| SMILES | OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 1149-16-2 | |
| 240.26 | |
| OMJNMRJMNUREOJ-KOBPNRPCSA-N | |
| OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O |
| C14H12N2O2 | |
| MFCD00002183 | |
| 5356911 |
Specifications
| 203°C to 211°C | |
| C14H12N2O2 | |
| NONH for all modes of transport | |
| OMJNMRJMNUREOJ-KOBPNRPCSA-N | |
| 6-[(E)-2-(2-hydroxyanilino)ethenyl]iminocyclohexa-2,4-dien-1-one | |
| 5356911 | |
| ≥97% (from N) | |
| Glass Bottle | |
| Yellow | |
| Glyoxal-bis(2-hydroxyanil) |
| 1149-16-2 | |
| MFCD00002183 | |
| 2808815 | |
| OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O | |
| 240.26 | |
| 240.26g/mol | |
| Odourless | |
| Passes test for suitable for metal definition | |
| 25 g |
Safety and Handling
RTECSNumber : 214-560-0