missing translation for 'onlineSavingsMsg'
Learn More

Glyoxal (39% in Water, ca. 8.8mol/L), TCI America™

Catalog No. G015225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G015225G 25 g
G0152500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G015225G Supplier TCI America Supplier No. G015225G

Chemical Identifiers

CAS 107-22-2
Molecular Formula C2H2O2
Molecular Weight (g/mol) 58.036
MDL Number MFCD00006957
InChI Key LEQAOMBKQFMDFZ-UHFFFAOYSA-N
Synonym glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40
PubChem CID 7860
ChEBI CHEBI:34779
IUPAC Name oxaldehyde
SMILES C(=O)C=O

Specifications

Physical Form Clear Liquid at 20°C
Color Yellow
pH 2.1 to 2.7
Quantity 25 g
Formula Weight 58.04
Chemical Name or Material Glyoxal (39% in Water, ca. 8.8mol/L)
EINECSNumber (2)-0510
RTECSNumber MD2700000
TSCA Yes