missing translation for 'onlineSavingsMsg'
Learn More

Glycyrrhizin 93.0+%, TCI America™

Catalog No. p-7130162
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01501G 1 g
G015025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G01501G Supplier TCI America Supplier No. G01501G

Chemical Identifiers

CAS 1405-86-3
Molecular Formula C42H62O16
Molecular Weight (g/mol) 822.94
MDL Number MFCD00065194,MFCD00065194
InChI Key LPLVUJXQOOQHMX-QWBHMCJMSA-N
Synonym Glycyrrhizic Acid
PubChem CID 134129369
IUPAC Name (2S,3S,4S,5R,6S)-6-{[(3S,4aR,6aR,6bS,8aS,11S,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-yl]oxy}-5-{[(2R,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxyoxan-2-yl]oxy}-3,4-dihydroxyoxane-2-carboxylic acid
SMILES CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O)O[C@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O)C(O)=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 822.94
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycyrrhizin
EINECSNumber (8)-0502&(9)-0398
RTECSNumber MD2025000
TSCA Yes