missing translation for 'onlineSavingsMsg'
Learn More

Glycyrrhizic acid monoammonium salt, 98+%

Catalog No. p-139275
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC213750100 10 g Glass bottle
AC213750250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC213750100 Supplier Thermo Scientific Chemicals Supplier No. 213750100
Only null left

CAS: 53956-04-0 | C42H65NO16 | 839.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53956-04-0
Molecular Formula C42H65NO16
Molecular Weight (g/mol) 839.97
MDL Number MFCD00167400,MFCD00167400
InChI Key ILRKKHJEINIICQ-OOFFSTKBSA-N
PubChem CID 131676200
IUPAC Name (2S,3S,4S,5R,6R)-6-[(2S,3R,4S,5S,6S)-2-[[(4aR,6aR,6bS,8aS,11R,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-2,3,4a,5,6,7,8,9,10,12,12a,14a-dodecahydro-1H-picen-3-yl]oxy]-6-carboxy-4,5-dihydroxyoxan-3-yl]oxy-3,4,5-trihydroxyoxane-2-carb
SMILES N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O)O[C@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O)C(O)=O

Specifications

Melting Point 209.0°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 10 g
Merck Index 10,4369
Specific Rotation Condition + 51.50 (20.00°C c=1,EtOH 50% v/v)
Specific Rotation + 51.50
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid, ethanol,, soluble in ammonia water
Formula Weight 839.98
Percent Purity 98+%
Related Substances 10% max. 18-alpha-glycyrrhizic acid (HPLC)
Physical Form Crystals or Powder
Chemical Name or Material Glycyrrhizic acid monoammonium salt, 98%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 258-887-7
RTECSNumber LZ6500000
TSCA TSCA

RUO – Research Use Only