missing translation for 'onlineSavingsMsg'
Learn More

Glycyrrhetic Acid 97.0+%, TCI America™

Catalog No. G01491G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01491G 1 g
G014925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G01491G Supplier TCI America Supplier No. G01491G

Chemical Identifiers

CAS 471-53-4
Molecular Formula C30H46O4
Molecular Weight (g/mol) 470.69
MDL Number MFCD00003706,MFCD00066716
InChI Key MPDGHEJMBKOTSU-YKLVYJNSSA-N
Synonym enoxolone, glycyrrhetinic acid, uralenic acid, glycyrrhetic acid, 18-beta-glycyrrhetinic acid, glycyrrhetin, arthrodont, jintan, 18beta-glycyrrhetinic acid, rhetinic acid
PubChem CID 10114
ChEBI CHEBI:30853
IUPAC Name (2S,4aS,6aS,6bR,8aR,10S,12aS,12bR,14bR)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-2-carboxylic acid
SMILES CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O

Specifications

Melting Point 297°C
Color White
Quantity 1 g
Formula Weight 470.69
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycyrrhetic Acid
EINECSNumber (8)-0507&(9)-0399
RTECSNumber RK0180000
TSCA Yes