missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycylglycine 99.0+%, TCI America™
Supplier: TCI America G0124100G
Specifications
| 260°C | |
| 556-50-3 | |
| C4H8N2O3 | |
| glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid | |
| C(C(=O)NCC(=O)O)N | |
| 100 g | |
| 11163 | |
| 132.12 | |
| Crystalline Powder |
| Glycylglycine | |
| White | |
| MFCD00008130 | |
| YMAWOPBAYDPSLA-UHFFFAOYSA-N | |
| 2-[(2-aminoacetyl)amino]acetic acid | |
| 132.119 | |
| CHEBI:17201 | |
| ≥99.0% (T) |
Chemical Identifiers
| 556-50-3 | |
| 132.119 | |
| YMAWOPBAYDPSLA-UHFFFAOYSA-N | |
| 11163 | |
| 2-[(2-aminoacetyl)amino]acetic acid |
| C4H8N2O3 | |
| MFCD00008130 | |
| glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid | |
| CHEBI:17201 | |
| C(C(=O)NCC(=O)O)N |
Safety and Handling
EINECSNumber : (2)-2667
TSCA : Yes