missing translation for 'onlineSavingsMsg'
Learn More

Glycylglycine 99.0+%, TCI America™

Catalog No. G012425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G012425G 25 g
G0124100G 100 g
G0124500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G012425G Supplier TCI America Supplier No. G012425G

Chemical Identifiers

CAS 556-50-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00008130
InChI Key YMAWOPBAYDPSLA-UHFFFAOYSA-N
Synonym glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid
PubChem CID 11163
ChEBI CHEBI:17201
IUPAC Name 2-[(2-aminoacetyl)amino]acetic acid
SMILES C(C(=O)NCC(=O)O)N

Specifications

Melting Point 260°C
Color White
Quantity 25 g
Formula Weight 132.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycylglycine
EINECSNumber (2)-2667
TSCA Yes