missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Glycylglycine, Free Base, Calbiochem™,

Catalog No. 3630100GM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3630100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3630100GM Supplier MilliporeSigma™ Supplier No. 3630100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity chemical reagent used in biochemical research.

Chemical Identifiers

CAS 556-50-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
InChI Key YMAWOPBAYDPSLA-UHFFFAOYSA-N
Synonym glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid
PubChem CID 11163
ChEBI CHEBI:17201
IUPAC Name 2-[(2-aminoacetyl)amino]acetic acid
SMILES C(C(=O)NCC(=O)O)N

Specifications

Color White
Packaging Glass Bottle
Quantity 100 g
Percent Purity >98% (TLC)
Grade Biochemical
Physical Form Solid
Chemical Name or Material Glycylglycine