missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycylglycine, 99%

Catalog No. AAA1052314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1052314 25 g
AAA1052322 100 g
AAA1052336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1052314 Supplier Thermo Scientific Chemicals Supplier No. A1052314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 556-50-3 | C4H8N2O3 | 132.119 g/mol

Glycylglycine is as a buffering reagent and also used in biochemical research. It is the simplest of all dipeptides and is used as a starting template for preparation of more complex peptides. It is also employed as a practical buffer with a buffering range of 7.5 - 8.9 (at 25°C). Valuable agent for experiments investigating peptide transport and absorption.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycylglycine is as a buffering reagent and also used in biochemical research. It is the simplest of all dipeptides and is used as a starting template for preparation of more complex peptides. It is also employed as a practical buffer with a buffering range of 7.5 - 8.9 (at 25°C). Valuable agent for experiments investigating peptide transport and absorption.

Solubility
Soluble in water (132 g/L) at 20°C, and ethanol (slightly solube).

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 556-50-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00008130
InChI Key YMAWOPBAYDPSLA-UHFFFAOYSA-N
Synonym glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid
PubChem CID 11163
ChEBI CHEBI:17201
IUPAC Name 2-[(2-aminoacetyl)amino]acetic acid
SMILES C(C(=O)NCC(=O)O)N

Specifications

Melting Point ∼220°C (decomposition)
Beilstein 1765223
Merck Index 14,4503
Solubility Information Soluble in water (132g/L) at 20°C,and ethanol (slightly solube).
Quantity 25 g
Formula Weight 132.12
Percent Purity 99%
Chemical Name or Material Glycylglycine
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 209-127-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only