missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Glycylglycine, 99+%
CAS: 556-50-3 | C4H8N2O3 | 132.12 g/mol
$100.92 - $270.95
Chemical Identifiers
| CAS | 556-50-3 |
|---|---|
| Molecular Formula | C4H8N2O3 |
| Molecular Weight (g/mol) | 132.12 |
| MDL Number | MFCD00008130 |
| InChI Key | YMAWOPBAYDPSLA-UHFFFAOYSA-N |
| Synonym | glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid |
| PubChem CID | 11163 |
| ChEBI | CHEBI:17201 |
| IUPAC Name | 2-[(2-aminoacetyl)amino]acetic acid |
| SMILES | C(C(=O)NCC(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC120140250
|
Thermo Scientific Chemicals
120140250 |
25 g |
Each for $100.92
|
|
|||||
|
AC120141000
|
Thermo Scientific Chemicals
120141000 |
100 g |
Each for $270.95
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
| 556-50-3 | |
| 132.12 | |
| YMAWOPBAYDPSLA-UHFFFAOYSA-N | |
| 11163 | |
| 2-[(2-aminoacetyl)amino]acetic acid |
| C4H8N2O3 | |
| MFCD00008130 | |
| glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid | |
| CHEBI:17201 | |
| C(C(=O)NCC(=O)O)N |
Specifications
| 262.0°C to 264.0°C | |
| 556-50-3 | |
| White | |
| Authentic | |
| 99+% | |
| Glass Bottle | |
| MFCD00008130 | |
| 15, 4539 | |
| glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid | |
| YMAWOPBAYDPSLA-UHFFFAOYSA-N | |
| 2-[(2-aminoacetyl)amino]acetic acid | |
| 25 g | |
| CHEBI:17201 | |
| ≥99% | |
| Glycylglycine, 99+% |
| (10% aq. soln.) (1 cm cell), 0.01 max. at 405nm | |
| 5.5 to 6 (5% aq. soln.) | |
| 100ppm max | |
| 0.5% max. (105°C, 4 hrs) | |
| C4H8N2O3 | |
| H2NCH2CONHCH2CO2H | |
| 04, 371 | |
| 0.1% max. | |
| Solubility in water: 132g/L (20°C). Other solubilities: slightly soluble in ethanol, practically insoluble in ether | |
| C(C(=O)NCC(=O)O)N | |
| 132.12 | |
| 11163 | |
| 132.12 | |
| Fine Crystalline Powder |
Safety and Handling
EINECSNumber : 209-127-8
TSCA : TSCA
RUO – Research Use Only