missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycylglycine, 99+%

Catalog No. AC120141000
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC120141000 100 g
AC120140250 25 g
AC120140010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC120141000 Supplier Thermo Scientific Chemicals Supplier No. 120141000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 556-50-3 | C4H8N2O3 | 132.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 556-50-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.12
MDL Number MFCD00008130
InChI Key YMAWOPBAYDPSLA-UHFFFAOYSA-N
Synonym glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid
PubChem CID 11163
ChEBI CHEBI:17201
IUPAC Name 2-[(2-aminoacetyl)amino]acetic acid
SMILES C(C(=O)NCC(=O)O)N

Specifications

Melting Point 262.0°C to 264.0°C
Absorbance (10% aq. soln.) (1 cm cell), 0.01 max. at 405nm
pH 5.5 to 6 (5% aq. soln.)
Color White
Chloride 100ppm max
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (105°C, 4 hrs)
Assay Percent Range 99+%
Packaging Plastic Bottle
Linear Formula H2NCH2CONHCH2CO2H
Beilstein 04, 371
Merck Index 15, 4539
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 132g/L (20°C). Other solubilities: slightly soluble in ethanol, practically insoluble in ether
Quantity 100 g
Formula Weight 132.12
Percent Purity ≥99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Glycylglycine, 99+%
Show More Show Less
EINECSNumber 209-127-8
TSCA TSCA

RUO – Research Use Only