missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycylglycine, ≥98.5%
CAS: 556-50-3 | C4H8N2O3 | 132.119 g/mol
$158.29 - $1184.14
Chemical Identifiers
| CAS | 556-50-3 |
|---|---|
| Molecular Formula | C4H8N2O3 |
| Molecular Weight (g/mol) | 132.119 |
| MDL Number | MFCD00008130 |
| InChI Key | YMAWOPBAYDPSLA-UHFFFAOYSA-N |
| Synonym | glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid |
| PubChem CID | 11163 |
| ChEBI | CHEBI:17201 |
| IUPAC Name | 2-[(2-aminoacetyl)amino]acetic acid |
| SMILES | C(C(=O)NCC(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AAJ1650522
|
Thermo Fisher Scientific
J1650522 |
100 g | Plastic Bottle |
Each for $158.29
|
|
||||
|
AAJ16505A1
|
Thermo Fisher Scientific
J16505A1 |
1 kg | Plastic Bottle |
Each for $1,184.14
|
|
||||
Chemical Identifiers
| 556-50-3 | |
| 132.119 | |
| YMAWOPBAYDPSLA-UHFFFAOYSA-N | |
| 11163 | |
| 2-[(2-aminoacetyl)amino]acetic acid |
| C4H8N2O3 | |
| MFCD00008130 | |
| glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid | |
| CHEBI:17201 | |
| C(C(=O)NCC(=O)O)N |
Specifications
| ∼220°C (decomposition) | |
| C4H8N2O3 | |
| NH2CH2CONHCH2COOH | |
| 1765223 | |
| YMAWOPBAYDPSLA-UHFFFAOYSA-N | |
| 2-[(2-aminoacetyl)amino]acetic acid | |
| 132.119 | |
| CHEBI:17201 | |
| ≥98.5% | |
| Glycylglycine |
| 556-50-3 | |
| Plastic Bottle | |
| MFCD00008130 | |
| glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid | |
| C(C(=O)NCC(=O)O)N | |
| 100 g | |
| 11163 | |
| 132.12 | |
| Crystalline Powder |
Safety and Handling
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 209-127-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only