missing translation for 'onlineSavingsMsg'
Learn More

Glycylglycine, ≥98.5%

Catalog No. AAJ1650522
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1650522 100 g Plastic Bottle
AAJ16505A1 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ1650522 Supplier Thermo Fisher Scientific Supplier No. J1650522

CAS: 556-50-3 | C4H8N2O3 | 132.119 g/mol

Chemical Identifiers

CAS 556-50-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00008130
InChI Key YMAWOPBAYDPSLA-UHFFFAOYSA-N
Synonym glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid
PubChem CID 11163
ChEBI CHEBI:17201
IUPAC Name 2-[(2-aminoacetyl)amino]acetic acid
SMILES C(C(=O)NCC(=O)O)N

Specifications

Melting Point ∼220°C (decomposition)
Packaging Plastic Bottle
Linear Formula NH2CH2CONHCH2COOH
Beilstein 1765223
Quantity 100 g
Formula Weight 132.12
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material Glycylglycine
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 209-127-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only