missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycyl-L-valine 99.0+%, TCI America™
$111.66 - $408.13
Chemical Identifiers
| CAS | 1963-21-9 |
|---|---|
| Molecular Formula | C7H14N2O3 |
| Molecular Weight (g/mol) | 174.2 |
| MDL Number | MFCD00066046 |
| InChI Key | STKYPAFSDFAEPH-LURJTMIESA-N |
| Synonym | glycyl-l-valine, h-gly-val-oh, gly-val, s-2-2-aminoacetamido-3-methylbutanoic acid, n-glycyl-l-valine, gly-l-val, valine, n-glycyl-, l, chembl56035, glycyl-valine, gly-d-val |
| PubChem CID | 2724807 |
| ChEBI | CHEBI:73922 |
| IUPAC Name | (2S)-2-[(2-aminoacetyl)amino]-3-methylbutanoic acid |
| SMILES | CC(C)C(C(=O)O)NC(=O)CN |
Chemical Identifiers
| 1963-21-9 | |
| 174.2 | |
| STKYPAFSDFAEPH-LURJTMIESA-N | |
| 2724807 | |
| (2S)-2-[(2-aminoacetyl)amino]-3-methylbutanoic acid |
| C7H14N2O3 | |
| MFCD00066046 | |
| glycyl-l-valine, h-gly-val-oh, gly-val, s-2-2-aminoacetamido-3-methylbutanoic acid, n-glycyl-l-valine, gly-l-val, valine, n-glycyl-, l, chembl56035, glycyl-valine, gly-d-val | |
| CHEBI:73922 | |
| CC(C)C(C(=O)O)NC(=O)CN |
Specifications
| 1963-21-9 | |
| C7H14N2O3 | |
| glycyl-l-valine, h-gly-val-oh, gly-val, s-2-2-aminoacetamido-3-methylbutanoic acid, n-glycyl-l-valine, gly-l-val, valine, n-glycyl-, l, chembl56035, glycyl-valine, gly-d-val | |
| CC(C)C(C(=O)O)NC(=O)CN | |
| 1 g | |
| 2724807 | |
| 174.20 | |
| Crystalline Powder |
| White | |
| MFCD00066046 | |
| STKYPAFSDFAEPH-LURJTMIESA-N | |
| (2S)-2-[(2-aminoacetyl)amino]-3-methylbutanoic acid | |
| 174.2 | |
| CHEBI:73922 | |
| ≥99.0% (T) | |
| Glycyl-L-valine |
Safety and Handling
TSCA : No