missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycyl-L-valine 99.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 174.20 |
| Percent Purity | ≥99.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Glycyl-L-valine |
Safety and Handling
| TSCA | No |