missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-tyrosine Hydrate 98.0+%, TCI America™

Catalog No. G01451G
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01451G 1g
G01455G 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G01451G Supplier TCI America Supplier No. G01451G
Only null left

Specifications

Purity ≥98.0% (T)
CAS 207300-83-2
Color White-Yellow
Format Crystal-Powder at 20°C
Molecular Weight (g/mol) 267.30
Product Type Glycyl-L-tyrosine Hydrate
Quantity 1g