missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycyl-DL-norleucine 98.0+%, TCI America™
$172.77 - $850.57
Chemical Identifiers
| CAS | 1504-41-2 |
|---|---|
| Molecular Formula | C8H16N2O3 |
| Molecular Weight (g/mol) | 188.227 |
| MDL Number | MFCD00038187 |
| InChI Key | XVUIZOUTLADVIH-UHFFFAOYSA-N |
| Synonym | H-Gly-DL-Nle-OH |
| PubChem CID | 273080 |
| IUPAC Name | 2-[(2-aminoacetyl)amino]hexanoic acid |
| SMILES | CCCCC(C(=O)O)NC(=O)CN |
Chemical Identifiers
| 1504-41-2 | |
| 188.227 | |
| XVUIZOUTLADVIH-UHFFFAOYSA-N | |
| 273080 | |
| CCCCC(C(=O)O)NC(=O)CN |
| C8H16N2O3 | |
| MFCD00038187 | |
| H-Gly-DL-Nle-OH | |
| 2-[(2-aminoacetyl)amino]hexanoic acid |
Specifications
| 227°C | |
| White | |
| MFCD00038187 | |
| XVUIZOUTLADVIH-UHFFFAOYSA-N | |
| 2-[(2-aminoacetyl)amino]hexanoic acid | |
| 188.227 | |
| 188.23 | |
| Crystalline Powder |
| 1504-41-2 | |
| C8H16N2O3 | |
| H-Gly-DL-Nle-OH | |
| CCCCC(C(=O)O)NC(=O)CN | |
| 1 g | |
| 273080 | |
| ≥98.0% (T) | |
| Glycyl-DL-norleucine |
Safety and Handling
TSCA : No