missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycyl-D-threonine Dihydrate 95.0+%, TCI America™
Specifications
Specifications
| Purity | ≥95.0% (T) |
| CAS | 74807-44-6 |
| Color | White |
| Format | Crystal-Powder at 20°C |
| Molecular Weight (g/mol) | 212.202 |
| Product Type | Glycyl-D-threonine Dihydrate |
| Quantity | 100 mg |