missing translation for 'onlineSavingsMsg'
Learn More

Glycopyrrolate 98.0+%, TCI America™

Catalog No. g0392100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0392100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G0392100MG Supplier TCI America Supplier No. G0392100MG
Only null left

Chemical Identifiers

CAS 596-51-0
Molecular Formula C19H28BrNO3
Molecular Weight (g/mol) 398.34
MDL Number MFCD00072137
InChI Key VPNYRYCIDCJBOM-UHFFFAOYNA-M
Synonym 3-(2-Cyclopentyl-2-hydroxy-2-phenylacetoxy)-1,1-dimethylpyrrolidinium Bromide
PubChem CID 11693
IUPAC Name 3-[(2-cyclopentyl-2-hydroxy-2-phenylacetyl)oxy]-1,1-dimethylpyrrolidin-1-ium bromide
SMILES [Br-].C[N+]1(C)CCC(C1)OC(=O)C(O)(C1CCCC1)C1=CC=CC=C1

Specifications

Melting Point 195°C
Color White
Quantity 100 mg
Formula Weight 398.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Glycopyrrolate
RTECSNumber UY4337630
TSCA No
Recommended Storage Refrigerator