missing translation for 'onlineSavingsMsg'
Learn More

Cuprous Cyanide, Powder, Purified, Spectrum™ Chemical

Catalog No. 18610072
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610072 45 kg Amber Glass Bottle
18610067 25 g Amber Glass Bottle
18610068 125 g Amber Glass Bottle
18610069 500 g Amber Glass Bottle
18610070 2.5 kg Amber Glass Bottle
18610071 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610072 Supplier Spectrum Chemical Mfg Cor Supplier No. C144545KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 544-92-3 | 89.56 g/mol

Chemical Identifiers

CAS 544-92-3

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 45 kg
Molecular Weight (g/mol) 89.56
Grade Technical
Packaging Amber Glass Bottle
Chemical Name or Material Cuprous Cyanide, Powder

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.