missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycolic Acid, Technical, 70-72%, Spectrum™ Chemical
GL185, C2H4O3
Supplier: Spectrum Chemical Mfg Cor GL1854LTGL
Description
Spectrum™ Chemical Glycolic Acid, 70 Percent, Technical , is slightly stronger than acetic acid. It is often included into emulsion polymers, solvents and additives for ink and paint in order to improve flow properties and impart gloss. It is used in surface treatment products that increase the coefficient of friction on tile flooring. Glycolic acid is a useful synthetic organic intermediate for oxidation-reduction, esterification and long chain polymerizations.Specifications
| 79-14-1,7732-18-5 | |
| C2H4O3 | |
| AEMRFAOFKBGASW-UHFFFAOYSA-N | |
| 2-hydroxyacetic acid | |
| 70 to 72% | |
| 4 L |
| 70%,30% | |
| UN3265 | |
| OCC(O)=O | |
| 76.05 | |
| Technical | |
| Amber Glass Bottle |
Chemical Identifiers
| 79-14-1 | |
| 76.05 | |
| 2-hydroxyacetic acid |
| C2H4O3 | |
| AEMRFAOFKBGASW-UHFFFAOYSA-N | |
| OCC(O)=O |