missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycolic Acid 98.0+%, TCI America™
Specifications
Specifications
| pH | 0.1 (70% H2O soln. 25°C) |
| Melting Point | 76°C |
| Color | White |
| Quantity | 25 g |
| Physical Form | Crystalline Powder |
| UN Number | 3261 |
| Formula Weight | 76.05 |
| Percent Purity | ≥98.0% (GC,T) |
| Chemical Name or Material | Glycolic Acid |
Safety and Handling
| EINECSNumber | (2)-1346 |
| RTECSNumber | MC5250000 |
| TSCA | Yes |